(4-Chlor-2-fluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Chloro-2-fluorophenylacetic acid

009422-1G 009422-5G
FLUO009422-1GEA 31.1 EUR
FLUO009422-1G FLUO009422-5G
(4-Chlor-2-fluorphenyl)essigsäure
(4-Chlor-2-fluorphenyl)essigsäure
Formel: (C₆H₃ClF)CH₂COOH
Molecular Weight: 188,59 g/mol
MDL: MFCD03095402
CAS-Nummer: 194240-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR