3,3-Diphenylpropionsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,3-Diphenylpropanoic acid

117420250 117421000
ACRO117420250EA 33.9 EUR
ACRO117420250 ACRO117421000
3,3-Diphenylpropionsäure 97%
3,3-Diphenylpropionsäure
Formel: C₁₅H₁₄O₂
Siedepunkt: 365 °C (1013 hPa)
Schmelzpunkt: 150…155 °C
Flammpunkt: 113 °C
MDL: MFCD00002717
CAS-Nummer: 606-83-7
EINECS: 210-125-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
GC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %
Iron (Fe) ≤10 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR