4-Phenylbuttersäure 99%
Lieferant: Thermo Scientific
Synonyme:
4-Phenylbutanoic acid
130380250
130381000
130385000
ACRO130380250EA
43
EUR
ACRO130380250
ACRO130381000
ACRO130385000
4-Phenylbuttersäure 99%
4-Phenylbuttersäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 165 °C (10 mmHg) Schmelzpunkt: 49…52 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004403 CAS-Nummer: 1821-12-1 EINECS: 217-341-8 |
Spezifikation Testergebnisse
Appearance | White to light yellow to beige Powder or crystals and/or chunks or fused solid |
Infrared spectrum | Conforms |
Melting point | 49 to 53 °C |
Titration with NaOH | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...