4-Phenylbuttersäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Phenylbutanoic acid
A18115.14
A18115.22
A18115.36
A18115.14EA
41.6
EUR
A18115.14
A18115.22
A18115.36
4-Phenylbuttersäure ≥99%
4-Phenylbuttersäure
Purity: 99%
CAS No.: 1821-12-1
Molecular Formula: C6H5(CH2)3CO2H
Formula Weight: 164.20
Flash Point:
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 165 °C (10 mmHg) Schmelzpunkt: 49…53 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004403 CAS-Nummer: 1821-12-1 EINECS: 217-341-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...