4-Phenylbuttersäure

Lieferant: Thermo Scientific

Synonyme: 4-Phenylbutanoic acid

SB02007.1 SB02007.10 SB02007.50
MAYBSB02007.1EA 0 EUR
MAYBSB02007.1 MAYBSB02007.10 MAYBSB02007.50
4-Phenylbuttersäure
4-Phenylbuttersäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
Siedepunkt: 290…294 °C (1013 hPa)
Schmelzpunkt: 48…51 °C
MDL: MFCD00004403
CAS-Nummer: 1821-12-1
EINECS: 217-341-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR