(4-Aminophenyl)essigsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Aminophenylacetic acid

104280250 104280050 104281000
ACRO104280250EA 60.8 EUR
ACRO104280250 ACRO104280050 ACRO104281000
(4-Aminophenyl)essigsäure 98%
(4-Aminophenyl)essigsäure
Formel: (C₆H₄(NH₂))CH₂COOH
Molecular Weight: 151.16 g/mol
Siedepunkt: 322
Schmelzpunkt: 196…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007916
CAS-Nummer: 1197-55-3
EINECS: 214-828-7
Merck Index: 13,00462

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 195 to 201 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.01 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.01 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.01 %
Strontium (Sr) ≤0.1 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 % (in dilute HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR