(4-Aminophenyl)essigsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Aminophenylacetic acid

A14675.09 A14675.18 A14675.30
A14675.09EA 33.2 EUR
A14675.09 A14675.18 A14675.30
(4-Aminophenyl)essigsäure 98%
(4-Aminophenyl)essigsäure

Purity: 98% CAS No.: 1197-55-3 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: ca 201° dec. Beilstein Reference: 2208094 Merck Reference: 14,463 MDL No.: MFCD00007916

Formel: (C₆H₄(NH₂))CH₂COOH
Molecular Weight: 151,16 g/mol
Siedepunkt: 322 °C (1013 hPa)
Schmelzpunkt: ∼ 201 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007916
CAS-Nummer: 1197-55-3
EINECS: 214-828-7
Merck Index: 14,463

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR