(2-Nitrophenyl)essigsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(2-Nitrophenyl)acetic acid
157060250
157061000
ACRO157060250EA
48.8
EUR
ACRO157060250
ACRO157061000
(2-Nitrophenyl)essigsäure 99%
(2-Nitrophenyl)essigsäure
Formel:
(C₆H₄NO₂)CH₂COOH Schmelzpunkt: 138…142 °C |
MDL:
MFCD00007190 CAS-Nummer: 3740-52-1 EINECS: 223-128-0 |
Spezifikation Testergebnisse
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 138 to 142 °C |
HPLC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...