(2-Nitrophenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(2-Nitrophenyl)acetic acid
M-Clarity™ Qualitätsstufe = MQ200
130249-100G
130249-25G
SIAL130249-100GEA
145
EUR
SIAL130249-100G
SIAL130249-25G
(2-Nitrophenyl)essigsäure, Sigma-Aldrich®
(2-Nitrophenyl)essigsäure
Formel:
(C₆H₄NO₂)CH₂COOH Molecular Weight: 181,15 g/mol Schmelzpunkt: 136…139 °C |
MDL:
MFCD00007190 CAS-Nummer: 3740-52-1 EINECS: 223-128-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...