(4-Nitrophenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrophenylacetic acid
M-Clarity™ Qualitätsstufe = MQ200
N20204-100G
N20204-25G
N20204-500G
SIALN20204-100GEA
90.3
EUR
SIALN20204-100G
SIALN20204-25G
SIALN20204-500G
(4-Nitrophenyl)essigsäure, Sigma-Aldrich®
(4-Nitrophenyl)essigsäure
Formel:
(C₆H₄(NO₂))CH₂COOH Molecular Weight: 181,15 g/mol Schmelzpunkt: 150…153 °C |
MDL:
MFCD00007383 CAS-Nummer: 104-03-0 EINECS: 203-168-5 Merck Index: 12,06719 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...