(2,4-Dinitrophenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

B22102.14 B22102.22
B22102.14EA 62.7 EUR
B22102.14 B22102.22
(2,4-Dinitrophenyl)essigsäure ≥98%
(2,4-Dinitrophenyl)essigsäure

Purity: 98% CAS No.: 643-43-6 Molecular Formula: C8H6N2O6 Formula Weight: 226.15 Melting Point: 177-182° Beilstein Reference: 1990798

Formel: (C₆H₃(NO₂)₂)CH₂COOH
Molecular Weight: 226,15 g/mol
Schmelzpunkt: 177…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007227
CAS-Nummer: 643-43-6
EINECS: 211-398-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR