(2-Methoxyphenyl)essigsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
177600250
177601000
ACRO177600250EA
59.2
EUR
ACRO177600250
ACRO177601000
(2-Methoxyphenyl)essigsäure 99%
(2-Methoxyphenyl)essigsäure
Formel:
(C₆H₄OCH₃)CH₂COOH Siedepunkt: 332 °C (1013 hPa) Schmelzpunkt: 122…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004321 CAS-Nummer: 93-25-4 EINECS: 202-231-4 |
Spezifikation Testergebnisse
Appearance | White to cream to light pink Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 122 to 125 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...