(2-Methoxyphenyl)essigsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Methoxyphenylacetic acid

A15211.09 A15211.14 A15211.18
A15211.09EA 27.8 EUR
A15211.09 A15211.14 A15211.18
(2-Methoxyphenyl)essigsäure ≥99%
(2-Methoxyphenyl)essigsäure

Purity: 99% CAS No.: 93-25-4 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 122-125° Beilstein Reference: 2047573

Formel: (C₆H₄OCH₃)CH₂COOH
Molecular Weight: 166.18 g/mol
Siedepunkt: 332
Schmelzpunkt: 122…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004321
CAS-Nummer: 93-25-4
EINECS: 202-231-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR