(3-Methoxyphenyl)essigsäure 99.5%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Methoxyphenylacetic acid
126010050
126010250
126011000
ACRO126010050EA
25.1
EUR
ACRO126010050
ACRO126010250
ACRO126011000
(3-Methoxyphenyl)essigsäure 99.5%
(3-Methoxyphenyl)essigsäure
Formel:
(C₆H₄(OCH₃))CH₂COOH Molecular Weight: 166.18 g/mol Siedepunkt: 305.95 Schmelzpunkt: 71…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004334 CAS-Nummer: 1798-09-0 EINECS: 217-282-8 |
Spezifikation Testergebnisse
Appearance | White to light beige Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
Titration with NaOH | ≥99.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...