(4-Benzyloxyphenyl)essigsäure

Lieferant: FLUOROCHEM

002506-5G 002506-25G
FLUO002506-5GEA 39.9 EUR
FLUO002506-5G FLUO002506-25G
(4-Benzyloxyphenyl)essigsäure
(4-Benzyloxyphenyl)essigsäure
Formel: (C₆H₅CH₂OC₆H₄)CH₂COOH
MDL: MFCD00017540
CAS-Nummer: 6547-53-1
EINECS: 229-463-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR