(4-Benzyloxyphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-(Benzyloxy)phenyl)acetic acid

M-Clarity™ Qualitätsstufe = MQ100

555398-1G
SIAL555398-1GEA 31 EUR
SIAL555398-1G
(4-Benzyloxyphenyl)essigsäure, Sigma-Aldrich®
(4-Benzyloxyphenyl)essigsäure
Formel: (C₆H₅CH₂OC₆H₄)CH₂COOH
Molecular Weight: 242.27 g/mol
MDL: MFCD00017540
CAS-Nummer: 6547-53-1
EINECS: 229-463-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR