(3-Hydroxy-4-methoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(3-Hydroxy-4-methoxyphenyl)acetic acid

002579-5G 002579-1G
FLUO002579-5GEA 152 EUR
FLUO002579-5G FLUO002579-1G
(3-Hydroxy-4-methoxyphenyl)essigsäure
(3-Hydroxy-4-methoxyphenyl)essigsäure
Formel: (C₆H₃(OH)(OCH₃))CH₂COOH
MDL: MFCD00016829
CAS-Nummer: 1131-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR