(3,4,5-Trimethoxyphenyl)essigsäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,4,5-Trimethoxyphenylacetic acid
B21136.06
B21136.14
B21136.22
B21136.06EA
42
EUR
B21136.06
B21136.14
B21136.22
(3,4,5-Trimethoxyphenyl)essigsäure ≥99%
(3,4,5-Trimethoxyphenyl)essigsäure
Purity: 99%
CAS No.: 951-82-6
Molecular Formula: C11H14O5
Formula Weight: 226.23
Melting Point: 118-122°
Beilstein Reference: 2697844
Formel:
(C₆H₂(OCH₃)₃)CH₂COOH Molecular Weight: 226,23 g/mol Siedepunkt: 352 °C (1013 hPa) Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004336 CAS-Nummer: 951-82-6 EINECS: 213-456-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...