(3,4,5-Trimethoxyphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALT70602-50GEA 208 EUR
SIALT70602-50G SIALT70602-10G
(3,4,5-Trimethoxyphenyl)essigsäure, Sigma-Aldrich®
(3,4,5-Trimethoxyphenyl)essigsäure
Formel: (C₆H₂(OCH₃)₃)CH₂COOH
Molecular Weight: 226,23 g/mol
Siedepunkt: 352 °C (1013 hPa)
Schmelzpunkt: 117…120 °C
MDL: MFCD00004336
CAS-Nummer: 951-82-6
EINECS: 213-456-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR