(3,5-Dimethoxyphenyl)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(3,5-Dimethoxyphenyl)acetic acid
B25672.03
B25672.06
B25672.03EA
49
EUR
B25672.03
B25672.06
(3,5-Dimethoxyphenyl)essigsäure ≥98%
(3,5-Dimethoxyphenyl)essigsäure
Purity: 98%
CAS No.: 4670-10-4
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 102-104°
MDL No.: MFCD00016827
Formel:
(C₆H₃(OCH₃)₂)CH₂COOH Molecular Weight: 196,2 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016827 CAS-Nummer: 4670-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...