(3-Hydroxyphenyl)essigsäure 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Hydroxyphenylacetic acid
121700050
121700250
ACRO121700050EA
54.8
EUR
ACRO121700050
ACRO121700250
(3-Hydroxyphenyl)essigsäure 99+%
(3-Hydroxyphenyl)essigsäure
Formel:
(C₆H₄(OH))CH₂COOH Molecular Weight: 152.15 g/mol Siedepunkt: 349 Schmelzpunkt: 128…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004337 CAS-Nummer: 621-37-4 EINECS: 210-684-4 |
Spezifikation Testergebnisse
Appearance | White to beige to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 128 to 131 °C |
HPLC | ≥99.0 % |
Titration with NaOH | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...