(3-Hydroxyphenyl)essigsäure ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Hydroxyphenylacetic acid

B23563.03 B23563.06 B23563.14
B23563.03EA 21.9 EUR
B23563.03 B23563.06 B23563.14
(3-Hydroxyphenyl)essigsäure ≥99%
(3-Hydroxyphenyl)essigsäure

Purity: 99% CAS No.: 621-37-4 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 128-132° Beilstein Reference: 2086506

Formel: (C₆H₄(OH))CH₂COOH
Molecular Weight: 152,15 g/mol
Siedepunkt: 349 °C (1013 hPa)
Schmelzpunkt: 128…132 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004337
CAS-Nummer: 621-37-4
EINECS: 210-684-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR