(3,4-Dihydroxyphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid

A15893.03 A15893.06 A15893.14
A15893.03EA 34.7 EUR
A15893.03 A15893.06 A15893.14
(3,4-Dihydroxyphenyl)essigsäure ≥98%
(3,4-Dihydroxyphenyl)essigsäure

Purity: 98+% CAS No.: 102-32-9 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 128-130° Beilstein Reference: 2211017

Formel: (C₆H₃(OH)₂)CH₂COOH
Molecular Weight: 168.15 g/mol
Siedepunkt: 337
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004338
CAS-Nummer: 102-32-9
EINECS: 203-024-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR