(3-Phenoxyphenyl)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Phenoxyphenylacetic acid
L12524.03
L12524.06
L12524.03EA
32.9
EUR
L12524.03
L12524.06
(3-Phenoxyphenyl)essigsäure ≥98%
(3-Phenoxyphenyl)essigsäure
Purity: 98%
CAS No.: 32852-81-6
Molecular Formula: C14H12O3
Formula Weight: 228.25
Melting Point: 88-90°
Beilstein Reference: 2730540
Formel:
(C₆H₅OC₆H₅)CH₂COOH Molecular Weight: 228,25 g/mol Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016826 CAS-Nummer: 32852-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...