(4-Phenoxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Phenoxyphenylacetic Acid

009338-1G 009338-5G
FLUO009338-1GEA 0 EUR
FLUO009338-1G FLUO009338-5G
(4-Phenoxyphenyl)essigsäure
(4-Phenoxyphenyl)essigsäure
Formel: (C₆H₅OC₆H₅)CH₂COOH
Molecular Weight: 228,25 g/mol
MDL: MFCD00079779
CAS-Nummer: 6328-74-1
UN: 3077
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR