(2,3,5-Trifluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-(2,3,5-Trifluorophenyl)acetic acid

007691-1G 007691-5G
FLUO007691-1GEA 381 EUR
FLUO007691-1G FLUO007691-5G
(2,3,5-Trifluorphenyl)essigsäure
(2,3,5-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Molecular Weight: 190.12 g/mol
MDL: MFCD00083547
CAS-Nummer: 132992-28-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR