(2,3,6-Trifluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-(2,3,6-Trifluorophenyl)acetic acid

007692-1G 007692-5G
FLUO007692-1GEA 45.9 EUR
FLUO007692-1G FLUO007692-5G
(2,3,6-Trifluorphenyl)essigsäure
(2,3,6-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Molecular Weight: 190,12 g/mol
MDL: MFCD00061217
CAS-Nummer: 114152-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR