(2,3-Difluorphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(2,3-Difluorophenyl)acetic acid

A16728.03 A16728.06
A16728.03EA 36.8 EUR
A16728.03 A16728.06
(2,3-Difluorphenyl)essigsäure ≥98%
(2,3-Difluorphenyl)essigsäure

Purity: 98% CAS No.: 145689-41-4 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 117-119° MDL No.: MFCD00040968

Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040968
CAS-Nummer: 145689-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR