(2,3-Difluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-(2,3-Difluorophenyl)acetic acid

001866-1G 001866-5G 001866-25G
FLUO001866-1GEA 31.1 EUR
FLUO001866-1G FLUO001866-5G FLUO001866-25G
(2,3-Difluorphenyl)essigsäure
(2,3-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
MDL: MFCD00040968
CAS-Nummer: 145689-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR