(2,4,5-Trifluorphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2,4,5-Trifluorophenylacetic acid

007693-1G 007693-5G 007693-25G
FLUO007693-1GEA 31.1 EUR
FLUO007693-1G FLUO007693-5G FLUO007693-25G
(2,4,5-Trifluorphenyl)essigsäure
(2,4,5-Trifluorphenyl)essigsäure
Formel: (C₆H₂F₃)CH₂COOH
Molecular Weight: 190.12 g/mol
MDL: MFCD00082479
CAS-Nummer: 209995-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR