(2,4,5-Trifluorphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,4,5-Trifluorophenylacetic acid
M-Clarity™ Qualitätsstufe = MQ100
684155-1G
684155-5G
SIAL684155-1GEA
33.7
EUR
SIAL684155-1G
SIAL684155-5G
(2,4,5-Trifluorphenyl)essigsäure, Sigma-Aldrich®
(2,4,5-Trifluorphenyl)essigsäure
Formel:
(C₆H₂F₃)CH₂COOH Molecular Weight: 190.12 g/mol |
MDL:
MFCD00082479 CAS-Nummer: 209995-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...