(2,4,6-Trifluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2,4,6-Trifluorophenylacetic acid

007694-1G 007694-5G
FLUO007694-1GEA 34.1 EUR
FLUO007694-1G FLUO007694-5G
(2,4,6-Trifluorphenyl)essigsäure
(2,4,6-Trifluorphenyl)essigsäure
Formel: (C₆H₂F₃)CH₂COOH
Molecular Weight: 190,12 g/mol
MDL: MFCD00061218
CAS-Nummer: 209991-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR