(2,4-Difluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2,4-Difluorophenylacetic acid

001927-5G 001927-25G
FLUO001927-5GEA 51.6 EUR
FLUO001927-5G FLUO001927-25G
(2,4-Difluorphenyl)essigsäure
(2,4-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172.13 g/mol
Siedepunkt: 259
Schmelzpunkt: 117…119 °C
MDL: MFCD00009999
CAS-Nummer: 81228-09-3
EINECS: 279-709-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR