(2,5-Difluorphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,5-Difluorophenylacetic acid

B21595.03 B21595.06 B21595.14
B21595.03EA 30.2 EUR
B21595.03 B21595.06 B21595.14
(2,5-Difluorphenyl)essigsäure ≥98%
(2,5-Difluorphenyl)essigsäure

Purity: 98% CAS No.: 85068-27-5 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 123-127° Beilstein Reference: 6844956

Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172.13 g/mol
Siedepunkt: 259
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010000
CAS-Nummer: 85068-27-5
EINECS: 285-288-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR