(2-Fluorphenyl)essigsäure
Lieferant: FLUOROCHEM
Synonyme:
2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid
001292-250G
001292-100G
001292-25G
FLUO001292-250GEA
203
EUR
FLUO001292-250G
FLUO001292-100G
FLUO001292-25G
(2-Fluorphenyl)essigsäure
(2-Fluorphenyl)essigsäure
Formel:
(C₆H₄F)CH₂COOH Siedepunkt: 263 °C (1013 hPa) Schmelzpunkt: 64 °C |
MDL:
MFCD00004315 CAS-Nummer: 451-82-1 EINECS: 207-196-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...