(3,4,5-Trifluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3,4,5-Trifluorophenylacetic acid

007695-1G 007695-5G
FLUO007695-1GEA 42.9 EUR
FLUO007695-1G FLUO007695-5G
(3,4,5-Trifluorphenyl)essigsäure
(3,4,5-Trifluorphenyl)essigsäure
Formel: (C₆H₂F₃)CH₂COOH
Molecular Weight: 190,12 g/mol
MDL: MFCD00083532
CAS-Nummer: 209991-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR