(3,5-Difluorphenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3,5-Difluorophenylacetic acid

002119-5G 002119-25G 002119-100G
FLUO002119-5GEA 31.1 EUR
FLUO002119-5G FLUO002119-25G FLUO002119-100G
(3,5-Difluorphenyl)essigsäure
(3,5-Difluorphenyl)essigsäure
Formel: (C₆H₃F₂)CH₂COOH
Molecular Weight: 172,13 g/mol
MDL: MFCD00010316
CAS-Nummer: 105184-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR