(4-Fluorphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(4-Fluorophenyl)acetic acid

001294-25G 001294-100G 001294-500G
FLUO001294-25GEA 30.8 EUR
FLUO001294-25G FLUO001294-100G FLUO001294-500G
(4-Fluorphenyl)essigsäure
(4-Fluorphenyl)essigsäure
Formel: (C₆H₄F)CH₂COOH
Molecular Weight: 154.14 g/mol
Siedepunkt: 264
Schmelzpunkt: 84…85 °C
MDL: MFCD00004343
CAS-Nummer: 405-50-5
EINECS: 206-972-4
Merck Index: 13,04204

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR