(2-(Trifluormethoxy)phenyl)essigsäure

Lieferant: CHEMPUR
Achtung

Synonyme: 2-(2-(Trifluoromethoxy)phenyl)acetic acid

FL7982.1 007982-5G 007982-25G
CHMPFL7982.1EA 0 EUR
CHMPFL7982.1 FLUO007982-5G FLUO007982-25G
(2-(Trifluormethoxy)phenyl)essigsäure
(2-(Trifluormethoxy)phenyl)essigsäure
Formel: F₃COC₆H₄CH₂COOH
Molecular Weight: 220,15 g/mol
MDL: MFCD00236325
CAS-Nummer: 220239-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR