(3-(Trifluormethoxy)phenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(Trifluoromethoxy)phenylacetic acid

002130-1G 002130-5G 002130-25G
FLUO002130-1GEA 0 EUR
FLUO002130-1G FLUO002130-5G FLUO002130-25G
(3-(Trifluormethoxy)phenyl)essigsäure
(3-(Trifluormethoxy)phenyl)essigsäure
Formel: (C₆H₄(OCF₃))CH₂COOH
Molecular Weight: 220.15 g/mol
MDL: MFCD00082480
CAS-Nummer: 203302-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR