(4-(Trifluormethoxy)phenyl)essigsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethoxy)phenylacetic acid
A19512.03
A19512.06
A19512.03EA
55.5
EUR
A19512.03
A19512.06
(4-(Trifluormethoxy)phenyl)essigsäure ≥98%
(4-(Trifluormethoxy)phenyl)essigsäure
Purity: 98%
CAS No.: 4315-07-5
Molecular Formula: C9H7F3O3
Formula Weight: 220.15
Melting Point: 86-88°
Beilstein Reference: 2695336
Formel:
(C₆H₄(OCF₃))CH₂COOH Molecular Weight: 220,15 g/mol Schmelzpunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061239 CAS-Nummer: 4315-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...