(4-(Trifluormethoxy)phenyl)essigsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-(Trifluoromethoxy)phenylacetic acid

002178-1G 002178-5G
FLUO002178-1GEA 31.1 EUR
FLUO002178-1G FLUO002178-5G
(4-(Trifluormethoxy)phenyl)essigsäure
(4-(Trifluormethoxy)phenyl)essigsäure
Formel: (C₆H₄(OCF₃))CH₂COOH
Molecular Weight: 220,15 g/mol
MDL: MFCD00061239
CAS-Nummer: 4315-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR