(2-Iodphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-(2-Iodophenyl)acetic acid
M-Clarity™ Qualitätsstufe = MQ200
531472-25G
531472-5G
SIAL531472-25GEA
92
EUR
SIAL531472-25G
SIAL531472-5G
(2-Iodphenyl)essigsäure, Sigma-Aldrich®
(2-Iodphenyl)essigsäure
Formel:
(C₆H₄I)CH₂COOH Molecular Weight: 262,05 g/mol |
MDL:
MFCD00046546 CAS-Nummer: 18698-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...