(3-Iodphenyl)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(3-Iodophenyl)acetic acid

L15117.04 L15117.ME
L15117.04EA 117 EUR
L15117.04 L15117.ME
(3-Iodphenyl)essigsäure ≥98%
(3-Iodphenyl)essigsäure

Purity: 98% CAS No.: 1878-69-9 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 128-132° Beilstein Reference: 2501867

Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262.05 g/mol
Schmelzpunkt: 128…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00046548
CAS-Nummer: 1878-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR