(3-Iodphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-(3-Iodophenyl)acetic acid

M-Clarity™ Qualitätsstufe = MQ100

587001-1G 587001-5G
SIAL587001-1GEA 33.7 EUR
SIAL587001-1G SIAL587001-5G
(3-Iodphenyl)essigsäure, Sigma-Aldrich®
(3-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262,05 g/mol
MDL: MFCD00046548
CAS-Nummer: 1878-69-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR