(4-Iodphenyl)essigsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(4-Iodophenyl)acetic acid
L13345.03
L13345.06
L13345.03EA
37.8
EUR
L13345.03
L13345.06
(4-Iodphenyl)essigsäure ≥97%
(4-Iodphenyl)essigsäure
Purity: 97%
CAS No.: 1798-06-7
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Melting Point: 136-140°
Beilstein Reference: 1940558
Formel:
(C₆H₄I)CH₂COOH Molecular Weight: 262.05 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082985 CAS-Nummer: 1798-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...