HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99%
Lieferant: Thermo Scientific
Synonyme: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
A14777.30 A14777.09 A14777.18
A14777.30EA 209 EUR
A14777.30 A14777.09 A14777.18
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99%
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)
Purity: 99% CAS No.: 7365-45-9 Molecular Formula: C8H18N2O4S Formula Weight: 238.31 Physical Form: Powder Appearance: White Melting Point: ca 236° dec. Beilstein Reference: 883043 Merck Reference: 14,4654 MDL No.: MFCD00006158
A zwitterionic buffer
Formel: C₈H₁₈N₂O₄S
Molecular Weight: 238,31 g/mol
Schmelzpunkt: ∼ 236 °C (Zersetzung)
Dichte: 1,448 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006158
CAS-Nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 14,4654
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM