HEPES Natriumsalz, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure Natriumsalz
391333-1
391333-100
391333-500
391333-1EA
864
EUR
391333-1
391333-100
391333-500
HEPES Natriumsalz, Millipore®
HEPES Natriumsalz
Formel:
C₈H₁₇N₂NaO₄S Molecular Weight: 260,29 g/mol Schmelzpunkt: ? 105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036463 CAS-Nummer: 75277-39-3 EINECS: 278-169-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...