trans-2-Octenal, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (E)-2-Octenal

M-Clarity™ Qualitätsstufe = MQ300

W321508-100G-K W321508-1KG-K
SIALW321508-100G-KEA 107 EUR
SIALW321508-100G-K SIALW321508-1KG-K
trans-2-Octenal, Sigma-Aldrich®
trans-2-Octenal
Formel: C₈H₁₄O
Molecular Weight: 126,2 g/mol
MDL: MFCD00007011
CAS-Nummer: 2548-87-0
EINECS: 219-833-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR