6-Chlor-1-hexen, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ100

543144-25G 543144-5G
SIAL543144-25GEA 188 EUR
SIAL543144-25G SIAL543144-5G
6-Chlor-1-hexen, Sigma-Aldrich®
6-Chlor-1-hexen
Formel: C₆H₁₁Cl
Molecular Weight: 118,61 g/mol
MDL: MFCD00039391
CAS-Nummer: 928-89-2
EINECS: 213-186-5
UN: 3295
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR